missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Nigericin sodium salt
GREENER_CHOICE

Product Code. 18722774
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18722774 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18722774 Supplier Tocris Bioscience™ Supplier No. 4312/10
  • ONLINE EXCLUSIVE PRICE
    145.00 €
    136.50 € / 10mg
    Save 8.50 €
    5% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Selective K+ ionophore

Potassium ionophore, exchanges K+ for H+ across biological membranes, in a similar manner to Valinomycin (Cat. No. 3373). Stimulates mitochondral ATPase activity and disrupts membrane potential. Also acts as an ionophore for Pb2+ with no activity with other divalent cations. Antibiotic derived from Streptomyces hygroscopius.
TRUSTED_SUSTAINABILITY

Specifications

CAS 28643-80-3
Quantity 10 mg
Molecular Formula C40H67NaO11
Formula Weight 746.94
Purity >98%
Molecular Weight (g/mol) 746.955
Chemical Name or Material Nigericin sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.