missing translation for 'onlineSavingsMsg'
Learn More

Nigericin Sodium Salt, TRC

Product Code. 30041177
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041177 10 mg 10mg
30040558 25 mg 25mg
30040921 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041177 Supplier Toronto Research Chemicals Supplier No. TRCN59852310MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28643-80-3
Molecular Formula C40H67NaO11
Molecular Weight (g/mol) 746.94
Synonym 1,6-Dioxaspiro[4.5]decane, Nigericin deriv., Antibiotic K 178, Antibiotic M 2, Antibiotic X 464, Azalomycin M, Helixin C, Polyetherin A, Stereoisomer of Tetrahydro-6-[[9-methoxy-2,4,10-trimethyl-2-[octahydro-2,3'-dimethyl-5'-[tetrahydro-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyl-2H-pyran-2-yl][2,2'-bifuran]-5-yl]-1,6-dioxaspiro[4.5]dec-7-yl]methyl]-α,3-dimethyl-2H-pyran-2-acetic Acid, X 464
IUPAC Name sodium;(2R)-2-[(2R,3S,6R)-6-[[(2S,4R,5R,6R,7R,9R)-2-[(2R,5S)-5-[(2R,3S,5R)-5-[(2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-3-methyloxolan-2-yl]-5-methyloxolan-2-yl]-7-methoxy-2,4,6-trimethyl-1,10-dioxaspiro[4.5]decan-9-yl]methyl]-3-methyloxan-2-yl]propanoate
SMILES C[C@H]1CC[C@H](C[C@@H]2C[C@H]([C@@H](C)[C@]3([C@H](C)C[C@@](C)([C@H]4CC[C@@](C)([C@H]5[C@@H](C)C[C@H]([C@@H]6[C@@H](C)C[C@@H](C)[C@@](CO)(O)O6)O5)O4)O3)O2)OC)O[C@H]1[C@@H](C)C(=O)[O-].[Na+]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C40H68O11.Na/c1-21-11-12-28(46-33(21)26(6)36(42)43)17-29-18-30(45-10)27(7)40(48-29)25(5)19-38(9,51-40)32-13-14-37(8,49-32)35-23(3)16-31(47-35)34-22(2)15-24(4)39(44,20-41)50-34;/h21-35,41,44H,11-20H2,1-10H3,(H,42,43);/q;+1/p-1/t21-,22-,23-,24+,25+,26+,27+,28+,29+,30+,31+,32+,33+,34-,35+,37-,38-,39-,40+;/m0./s1
Formula Weight 746.4581
Percent Purity >95%
Chemical Name or Material Nigericin Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.