missing translation for 'onlineSavingsMsg'
Learn More

Nigrosin water soluble

Product Code. 11483657
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11483657

Brand: Thermo Scientific Alfa Aesar A18147.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 8005-03-6 | C22H14N6Na2O9S2 | 616.49 g/mol

It is employed as a common biological stain for tissues and cells. Used to make inks and shoe-polish and for dyeing leather, wood, textiles, and plastics. The crude mixture of nigrosine hydrochlorides (Phenol Black L) is used to dye phenolic resins. Bases are used to make printing colors and to dye plastics. It finds its use in Dorner?s spore stain, as a negative stain in place of India ink for spirochetes, bacteria, protozoa and fungi. It is also used in compounding resins include phenolic resins, styrenics, polyamides and urea resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a common biological stain for tissues and cells. Used to make inks and shoe-polish and for dyeing leather, wood, textiles, and plastics. The crude mixture of nigrosine hydrochlorides (Phenol Black L) is used to dye phenolic resins. Bases are used to make printing colors and to dye plastics. It finds its use in Dorner′s spore stain, as a negative stain in place of India ink for spirochetes, bacteria, protozoa and fungi. It is also used in compounding resins include phenolic resins, styrenics, polyamides and urea resins.

Solubility
Soluble in water, ethanol, oil, nigrosines and fat.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8005-03-6
Molecular Formula C22H14N6Na2O9S2
Molecular Weight (g/mol) 616.49
MDL Number MFCD00044681
InChI Key HKBVRFLHNUEVRO-HMDKGIQMSA-L
Synonym Acid Black 2, C.I. 50420
PubChem CID 44134531
IUPAC Name disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Quantity 100 g
Chemical Name or Material Nigrosin, water soluble

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.