missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nigrosine, pure, water soluble

Product Code. 10177370 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10177370

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 8005-03-6 | C22H14N6Na2O9S2 | 616.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8005-03-6
Molecular Formula C22H14N6Na2O9S2
Molecular Weight (g/mol) 616.49
MDL Number MFCD00044681
InChI Key HKBVRFLHNUEVRO-HMDKGIQMSA-L
Synonym Acid Black 2, C.I. 50420
PubChem CID 44134531
IUPAC Name disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Black to Black/Red
Quantity 100 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Nigrosine, pract.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.