missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nile Blue A, pure, certified

Product Code. 10791822 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10791822 10 g Glass bottle 10g
10606124 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10791822 Supplier Thermo Scientific Chemicals Supplier No. 415690100
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3625-57-8 | C40H40N6O6S | 732.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3625-57-8
Molecular Formula C40H40N6O6S
Molecular Weight (g/mol) 732.84
MDL Number MFCD00064529
InChI Key QIRDPEPUXNCOLD-UHFFFAOYSA-N
Synonym Nile blue sulfate, C.I. 51180
PubChem CID 19256
IUPAC Name [9-(diethylamino)benzo[a]phenoxazin-5-ylidene]azanium;sulfate
SMILES CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.[O-]S(=O)(=O)[O-]

Specifications

Absorption Ratio 1.01 to 1.16
Dye Content 70% min.
Infrared Spectrum Authentic
Lambda Max 635 to 646nm
Packaging Glass bottle
Beilstein 27, 404
Color Black to Green
Quantity 10 g
Physical Form Powder
Formula Weight 732.87
Grade Pure
Chemical Name or Material Nile Blue A, Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.