missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nile Blue A

Product Code. 11493387
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493387 25 g 25g
11483387 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493387 Supplier Thermo Scientific Chemicals Supplier No. A17174.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3625-57-8 | C40H40N6O6S | 732.84 g/mol

Used in the preparation of an amperometric glucose sensor. Also can be employed in Lillie and Fullmer method for staining and discriminating between melanins and lipofuscins in paraffin sections of animal tissue; Lillie and Fuller method for fatty acids/lipids using frozen sections of fatty liver and avocado.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of an amperometric glucose sensor. Also can be employed in Lillie and Fullmer method for staining and discriminating between melanins and lipofuscins in paraffin sections of animal tissue; Lillie and Fuller method for fatty acids/lipids using frozen sections of fatty liver and avocado.

Solubility
Soluble in ethanol, and water (50 g/L) at 25°C.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3625-57-8
Molecular Formula C40H40N6O6S
Molecular Weight (g/mol) 732.84
MDL Number MFCD00064529
InChI Key QIRDPEPUXNCOLD-UHFFFAOYSA-N
PubChem CID 19256
IUPAC Name [9-(diethylamino)benzo[a]phenoxazin-5-ylidene]azanium;sulfate
SMILES CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.[O-]S(=O)(=O)[O-]

Specifications

Melting Point >300°C (decomposition)
Synonym C.I. 51180; Basic Blue 12
Solubility Information Soluble in ethanol,and water (50g/L) at 25°C.
Quantity 25 g
Formula Weight 732.84
Chemical Name or Material Nile Blue A

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.