missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Nile Blue A sulfate

Product Code. 16464716
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16464716 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16464716 Supplier MedChemExpress Supplier No. HY101900
 Restricted Item
Estimated Shipment: 24-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nile Blue A (Nile blue sulfate) is used to differentiate melanins and lipofuscins. It is also useful for staining fats and preparation of an amperometric glucose sensor.

Specifications

Quantity 100 mg
Storage Requirements 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Solubility DMSO : ≥ 150 mg/mL (409.37 mM) ∣ H2O : < 0.1 mg/mL (ultrasonic;warming) (insoluble)
Shipping Condition Room temperature
CAS 3625-57-8
Molecular Weight (g/mol) 366.42 g/mol
Synonym Nile blue sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.