missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Nile Red

Product Code. 16482316
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482316 500 mg 500mg
16494667 50 mg 50mg
16458896 100 mg 100mg
16438758 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16482316 Supplier MedChemExpress Supplier No. HYD0718
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Nile Red (Nile Blue A oxazone) is a selective and hydrophobic fluorescent stain for intracellular lipid droplets and neutral lipids.

Nile Red is intensely fluorescent in all organic solvents and the fluorescence colors range from golden yellow to deep red.

Specifications

Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Product Type Dye Reagents
Quantity 500 mg
Shipping Condition Room temperature
CAS 7385-67-3
Molecular Weight (g/mol) 318.37 g/mol
Synonym Nile Blue A oxazone ∣ Phenoxazone 9
Target HBV
Solubility DMSO : ≥ 50 mg/mL (157.05 mM) ∣ H2O : 0.1 mg/mL (0.31 mM; Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.