missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Nile Red

Product Code. 18709366
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18709366 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18709366 Supplier Tocris Bioscience™ Supplier No. 7387/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fluorogenic lipid membrane dye

Nile red is a fluorogenic lipid membrane dye, which fluoresces in the hydrophobic environment of lipid membranes. Non-fluorescent in polar environments (e.g. water). Nile red is suitable for long-term monitoring of lipid dynamics in living cells. Excitation maximum = 552 nm; emission maximum = 636 nm in methanol.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9-(Diethylamino)-5 H-benzo[a]phenoxazin-5-one
CAS 7385-67-3
Quantity 10 mg
Target Fluorescent Lipids Probes and Cell Membrane Stains
Molecular Formula C20H18N2O2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.