missing translation for 'onlineSavingsMsg'
Learn More

Nile Red Fluorescent stain for intracellular lipid droplets. MP Biomedicals

Product Code. 11472502 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11472502 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11472502 Supplier MP Biomedicals Supplier No. 0215174450
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7385-67-3
Molecular Formula C20H18N2O2
MDL Number MFCD00011639
Synonym Nile Blue A Oxazone, 9-diethylamino-5H-benzo-alpha-phenoxazine-5-one
InChI Key VOFUROIFQGPCGE-UHFFFAOYSA-N
SMILES CCN(CC)C1=CC=C2N=C3C(OC2=C1)=CC(=O)C1=CC=CC=C31
IUPAC Name 9-(diethylamino)benzo[a]phenoxazin-5-one
Molecular Weight (g/mol) 318.38
PubChem CID 65182
ChEBI CHEBI:52169
Formula Weight 318.4
Quantity 50 mg
Chemical Name or Material Nile Red, Fluorescent stain for intracellular lipid droplets.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.