missing translation for 'onlineSavingsMsg'
Learn More

Nilotinib monohydrochloride monohydrate, MedChemExpress

Product Code. 16444656
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16444656 100 mg 100mg
16440936 200 mg 200mg
16421536 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16444656 Supplier Medchem Express Supplier No. HY10159A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nilotinib monohydrochloride monohydrate is a second generation tyrosine kinase inhibitor (TKI), is significantly potent against BCR-ABL, and is active against many BCR-ABL mutants.

Nilotinib monohydrochloride monohydrate is a second generation tyrosine kinase inhibitor (TKI), is significantly potent against BCR-ABL, and is active against many BCR-ABL mutants.

Specifications

Chemical Name or Material Nilotinib monohydrochloride monohydrate
CAS 923288-90-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C28H25ClF3N7O2
Quantity 100 mg
Synonym AMN107 monohydrochloride monohydrate
Solubility Information DMSO : ≥ 33 mg/mL (56.51 mM) ∣H2O : 0.1 mg/mL (0.17 mM; Need ultrasonic)
SMILES O=C(NC1=CC(C(F)(F)F)=CC(N2C=NC(C)=C2)=C1)C3=CC=C(C)C(NC4=NC=CC(C5=CC=CN=C5)=N4)=C3.[H]Cl.O
Molecular Weight (g/mol) 583.99
Formula Weight 583.99
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.