missing translation for 'onlineSavingsMsg'
Learn More

Nilotinib Monohydrochloride Monohydrate, TRC

Product Code. 30484277
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30484277 5 g 5g
30466396 100 mg -
30484163 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30484277 Supplier Toronto Research Chemicals Supplier No. TRCN4652955G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 923288-90-8
Molecular Formula C28 H22 F3 N7 O . Cl H . H2 O
Molecular Weight (g/mol) 583.99
Synonym Benzamide, 4-methyl-N-[3-(4-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]-, hydrochloride, hydrate (1:1:1), Nilotinib monohydrochloride monohydrate
IUPAC Name 4-methyl-N-[3-(4-methylimidazol-1-yl)-5-(trifluoromethyl)phenyl]-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]benzamide;hydrate;hydrochloride
SMILES O.Cl.Cc1cn(cn1)c2cc(NC(=O)c3ccc(C)c(Nc4nccc(n4)c5cccnc5)c3)cc(c2)C(F)(F)F

Specifications

Quantity 5 g
InChI Formula InChI=1S/C28H22F3N7O.ClH.H2O/c1-17-5-6-19(10-25(17)37-27-33-9-7-24(36-27)20-4-3-8-32-14-20)26(39)35-22-11-21(28(29,30)31)12-23(13-22)38-15-18(2)34-16-38;;/h3-16H,1-2H3,(H,35,39)(H,33,36,37);1H;1H2
Formula Weight 583.171
Chemical Name or Material Nilotinib Hydrochloride Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.