missing translation for 'onlineSavingsMsg'
Learn More

Nimodipine, MedChemExpress

Product Code. 16488196
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488196 100 mg 100mg
16462076 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488196 Supplier Medchem Express Supplier No. HYB0265
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nimodipine (BAY-e 9736) is an orally active, well-tolerated and light-sensitive dihydropyridine calcium antagonist. Nimodipine can be used for the research of cerebrovascular disorders.

Nimodipine (BAY-e 9736) is an orally active, well-tolerated and light-sensitive dihydropyridine calcium antagonist. Nimodipine can be used for the research of cerebrovascular disorders.

Specifications

Chemical Name or Material Nimodipine
CAS 66085-59-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H26N2O7
Quantity 100 mg
Synonym BAY-e 9736
Solubility Information DMSO : 100 mg/mL (238.98 mM; Need ultrasonic)
SMILES O=C(C1=C(C)NC(C)=C(C(OC(C)C)=O)C1C2=CC=CC([N+]([O-])=O)=C2)OCCOC
Molecular Weight (g/mol) 418.44
Formula Weight 418.44
Percent Purity 98.71%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.