missing translation for 'onlineSavingsMsg'
Learn More

Ninhydrin, 99%

Product Code. 11478603
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
250 g
Unit Size:
10g
250g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478603 250 g 250g
11458603 10 g 10g
11488730 25 g 25g
11468603 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11478603 Supplier Thermo Scientific Alfa Aesar Supplier No. A10409.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 485-47-2 | C9H6O4 | 178.143 g/mol

Ninhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ninhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.

Solubility
Soluble in water, acetone, ethanol, and methanol. Slightly soluble in chloroform and ether.

Notes
Light Sensitive. Incompatible with strong oxidizing agents, amines, alkalis and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 485-47-2
Molecular Formula C9H6O4
Molecular Weight (g/mol) 178.143
MDL Number MFCD00003791
InChI Key FEMOMIGRRWSMCU-UHFFFAOYSA-N
Synonym ninhydrin, ninhydrin hydrate, 1,2,3-indantrione monohydrate, ninhydrine, 2,2-dihydroxy-1h-indene-1,3 2h-dione, 2,2-dihydroxy-1,3-indandione, triketohydrindene hydrate, 1h-indene-1,3 2h-dione, 2,2-dihydroxy, 1,2,3-indantrione, 2-hydrate, 2,2-dihydroxyindane-1,3-dione
PubChem CID 10236
ChEBI CHEBI:86374
IUPAC Name 2,2-dihydroxyindene-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O

Specifications

Melting Point 251°C (decomposition)
Assay Percent Range 99%
Quantity 250 g
Beilstein 1910963
Sensitivity Light sensitive
Merck Index 14,6554
Solubility Information Soluble in water,acetone,ethanol,and methanol. Slightly soluble in chloroform and ether.
Formula Weight 178.14
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Ninhydrin
Show More Show Less
compliance-icons
Product Identifier
  • Ninhydrin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Ninhydrin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.