missing translation for 'onlineSavingsMsg'
Learn More

Ninhydrin, 0.35% (w/v) in Isopropyl Alcohol, Ricca Chemical

Product Code. 31940514
Change view
Click to view available options
Quantity:
120 mL
Unit Size:
120mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31940514 120 mL 120mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31940514 Supplier Ricca Chemical Company Supplier No. R5309350120C
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 485-47-2, 67-63-0
Packaging Amber Glass Bottle
Molecular Formula C9H6O4
Color Colorless to Yellow
Quantity 120 mL
InChI Key FEMOMIGRRWSMCU-UHFFFAOYSA-N
Physical Form Liquid
SMILES C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
IUPAC Name 2,2-dihydroxyindene-1,3-dione
Molecular Weight (g/mol) Mixture
PubChem CID 10236
ChEBI CHEBI:86374
Percent Purity 0.5% W/V
Grade Indicator
Chemical Name or Material Ninhydrin Solution 0.35% (w/v) in Isopropyl Alcohol, In Isopropanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.