missing translation for 'onlineSavingsMsg'
Learn More

Ninhydrin Spray Refill, Pure, 0.5% Ninhydrin In Butan-1-ol, Fisher Chemical

Product Code. 10254010 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10254010 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10254010 Supplier Fisher Chemical Supplier No. N/2161/90
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

500mL, Amber glass bottle

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -90°C
Boiling Point 117.4°C
CAS 485-47-2
Molecular Formula C9H6O4
MDL Number 3791
InChI Key FEMOMIGRRWSMCU-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
IUPAC Name 2,2-dihydroxyindene-1,3-dione
Molecular Weight (g/mol) 178.143
PubChem CID 10236
ChEBI CHEBI:86374
Formula Weight 74.0694g/mol
Packaging Amber glass bottle
Vapor Pressure 6.5&25
Color Colorless
Quantity 500 mL
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ninhydrin solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • Flammable liquid Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: 1H-Indene-1, 3(2H)-dione, 2, 2- dihydroxy- 485-47-2, n-Butylalcohol 71-36-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.