missing translation for 'onlineSavingsMsg'
Learn More

Ninhydrin, TRC

Product Code. 30479333
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479333 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30479333 Supplier Toronto Research Chemicals Supplier No. TRCN47828025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ninhydrin
CAS 485-47-2
Purity Grade Notes HPLC
Molecular Formula C9 H6 O4
Quantity 25 g
Synonym 2,2-Dihydroxy-1,3-indandione, 1H-Indene-1,2,3-trione 2-Hydrate, 2,2-Dihydroxy-1,3-dioxoindan, 2,2-Dihydroxy-1,3-indandione, 2,2-Dihydroxy-1H-indene-1,3(2H)-dione, 2,2-Dihydroxy-indene-1,3-dione, 2,2-Dihydroxyindane-1,3-dione, 2,3-Dihydro-2,2-dihydroxy-1,3-dioxo-1H-indene, Indane-1,2,3-trione monohydrate, Triketohydrindene Hydrate
InChI Formula InChI=1S/C9H6O4/c10-7-5-3-1-2-4-6(5)8(11)9(7,12)13/h1-4,12-13H
SMILES OC1(O)C(=O)c2ccccc2C1=O
IUPAC Name 2,2-dihydroxyindene-1,3-dione
Molecular Weight (g/mol) 178.14
Formula Weight 178.0266
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.