missing translation for 'onlineSavingsMsg'
Learn More

Skytec Ninja Lite Work Gloves

Product Code. 17892164 Shop All Skytec Products
Change view
Click to view available options
Size:
6, X-Small
7, Small
8, Medium
9, Large
10, X-Large
11, 2X-Large
12, 3X-Large
Unit Size:
10 pairs
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17892164 6, X-Small 10 pairs
17812174 7, Small 10 pairs
17802174 8, Medium 10 pairs
17822204 9, Large 10 pairs
17852164 10, X-Large 10 pairs
17882164 11, 2X-Large 10 pairs
17812164 12, 3X-Large 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 17892164 Supplier Skytec Supplier No. SKY120
  • ONLINE EXCLUSIVE PRICE
    50.90 €
    34.38 € / 10 pairs
    Save 16.52 €
    32% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ultra lightweight (18 G) Ninja Lite Work Gloves provide excellent second skin comfort as well as outstanding flexibility and a tactile touch. Protect the hand from oils, grease and abrasion, with effective grip. Ideal for intricate tasks and for preventing product contamination.

  • An ultra-lightweight glove, with exceptional flexibility and tactile touch for the most delicate of tasks
  • Superfine extra soft nylon liner
  • Maximum flexibility and movement for the most intricate of tasks
  • Breathable through front and back of the hand to reduce perspiration and skin irritation
  • Sanitized™ antimicrobial finish provides extra freshness

Specifications

Certifications/Compliance CAT II
Certifications/Compliance (Europe) EN388, EN420
Chlorinated No
Coated Area Palm
Construction Knit
Coating Material Polyurethane
Color Blue, Black
Exterior Finish No Texture
Hand Specific
Latex Free No
Material Nylon, Polyurethane
Product Type Work Glove
Size 6, X-Small
For Use With (Application) Fine handling
Glove Cuff Style Elasticated
Liner Material Nylon
Series Ninja Lite
Thumb Style Straight
Length (English) 9 in.
Length (Metric) 240 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.