missing translation for 'onlineSavingsMsg'
Learn More

Niobium Pentachloride, TRC

Product Code. 30466233
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity unitSize
30466233 50 g 50g
1 options
This item is not returnable. View return policy

Product Code. 30466233

Brand: Toronto Research Chemicals TRCN59853550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Niobium Pentachloride
CAS 10026-12-7
Molecular Formula Cl5Nb
Quantity 50 g
Synonym Niobium Chloride, Niobium Pentachloride, Niobium Pentachloride (NbCl5), Niobium(V) Chloride, Pentachloroniobium
InChI Formula InChI=1S/5ClH.Nd/h5*1H;/q;;;;;+5/p-5
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Nd+5]
IUPAC Name neodymium(5+);pentachloride
Molecular Weight (g/mol) 270.17
Formula Weight 318.7543
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.