missing translation for 'onlineSavingsMsg'
Learn More

Niobium(V) oxide, 99.95% (metals basis)

Product Code. 11357938
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357938 25 g 25g
11347938 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357938 Supplier Thermo Scientific Alfa Aesar Supplier No. 036478.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Vacuum Deposition | CAS: 1313-96-8 | Nb2O5 | 265.81 g/mol

Niobium(V) oxide is used for vacuum deposition, as an alternate electrode to lithium metal in advanced fuel cells, Applied in the solid state formation of an unusual cation deficient perovskite. And it is also used in Electrical and Electronic Products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-96-8
Molecular Formula Nb2O5
Molecular Weight (g/mol) 265.81
MDL Number MFCD00011128
InChI Key ZKATWMILCYLAPD-UHFFFAOYSA-N
Synonym niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde
PubChem CID 123105
SMILES O=[Nb](=O)O[Nb](=O)=O

Specifications

Melting Point 1520°C
Color White
Physical Form Sintered Lump
Quantity 25 g
Assay Percent Range 99.95% (metals basis)
Solubility Information Soluble in HF. Insoluble in water.
IUPAC Name [(dioxoniobio)oxy]dioxoniobium
Formula Weight 265.81
Odor Odorless
Appearance White 3-12mm Sintered Lump
Density 4.6
Chemical Name or Material Niobium(V) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.