missing translation for 'onlineSavingsMsg'
Learn More

Niobium(V) oxide, 99.99%, (trace metal basis)

Product Code. 10229693 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10229693 250 g Plastic bottle 250g
10785853 10 g Glass bottle 10g
10153970 50 g Plastic bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10229693 Supplier Thermo Scientific Acros Supplier No. 193632500
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1313-96-8 | Nb2O5 | 265.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-96-8
Molecular Formula Nb2O5
Molecular Weight (g/mol) 265.81
MDL Number MFCD00011128
InChI Key ZKATWMILCYLAPD-UHFFFAOYSA-N
Synonym niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde
PubChem CID 123105
SMILES O=[Nb](=O)O[Nb](=O)=O

Specifications

Melting Point 1520.0°C
Color White
CAS Min % 99.95
CAS Max % 100.0
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.99%
Linear Formula Nb2O5
Merck Index 15, 6645
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hf and hot h2so4,soluble in fused potassium hydrogen sulfate or,carbonates or hydroxides of the alkali metals
IUPAC Name [(dioxoniobio)oxy]dioxoniobium
Formula Weight 265.8
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Plastic bottle
Specific Gravity 4.47
Density 4.4700g/mL
Chemical Name or Material Niobium(V) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.