missing translation for 'onlineSavingsMsg'
Learn More

Nisoldipine, MedChemExpress

Product Code. 16447286
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16447286 100 mg 100mg
16411679 1 g 1g
16401856 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16447286 Supplier Medchem Express Supplier No. HY17402
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nisoldipine(BAY-k 5552; Sular) is a calcium channel blocker belonging to the dihydropyridines class, specific for L-type Cav1.2 with IC50 of 10 nM.

Nisoldipine(BAY-k 5552

Specifications

Chemical Name or Material Nisoldipine
CAS 63675-72-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H24N2O6
Quantity 100 mg
Synonym BAY-k 5552
Solubility Information DMSO : ≥ 100 mg/mL (257.46 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=C(C)NC(C)=C(C(OCC(C)C)=O)C1C2=CC=CC=C2[N+]([O-])=O)OC
Molecular Weight (g/mol) 388.41
Formula Weight 388.41
Percent Purity 98.22%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.