missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ NITD008

Product Code. 18703999
Change view
Click to view available options
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18703999 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18703999 Supplier R&D Systems™ Supplier No. 6045/1
  • ONLINE EXCLUSIVE PRICE
    231.22 €
    207.90 € / 1mg
    Save 23.32 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Antiviral; selective flavivirus inhibitor

NITD 008 is a selective flavivirus inhibitor. Adenosine analog. Inhibits dengue virus (EC50 = 0.64 μM), hepatitis C virus and cellular entry of Zika virus. Reduces increases of cytokines and NS1 and prevents death in dengue-infected mice. Active in vivo. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-(2-C-Ethynyl-β-D-ribofuranosyl)-7 H-pyrrolo[2,3-d]pyrimidin-4-amine
CAS 1044589-82-3
Assay Percent Range >98%
Quantity 1 mg
Product Line Tocris
Target Antiviral Compounds
Molecular Formula C13H14N4O4
Formula Weight Observed MW: 290.27
Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.