missing translation for 'onlineSavingsMsg'
Learn More

Nitenpyram, TRC

Product Code. 17912835
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17912835 1 g 1g
17920485 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17912835 Supplier Toronto Research Chemicals Supplier No. TRCN4901151G
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150824-47-8
Molecular Formula C11 H15 Cl N4 O2
Molecular Weight (g/mol) 270.72
Synonym 1,1-Ethenediamine, N-[(6-chloro-3-pyridinyl)methyl]-N-ethyl-N'-methyl-2-nitro-, (1E)-, 1,1-Ethenediamine, N-[(6-chloro-3-pyridinyl)methyl]-N-ethyl-N'-methyl-2-nitro-, (E)-, (1E)-N-[(6-Chloro-3-pyridinyl)methyl]-N-ethyl-N'-methyl-2-nitro-1,1-ethenediamine, (E)-N-(6-Chloro-3-pyridylmethyl)-N-ethyl-N'-methyl-2-nitroethylene-1,1-diamine, (E)-Nitenpyram, Bestguard, Capstar, Nitenpyram, Niterndipoine, TI 304
IUPAC Name (E)-1-N'-[(6-chloropyridin-3-yl)methyl]-1-N'-ethyl-1-N-methyl-2-nitroethene-1,1-diamine
SMILES CCN(Cc1ccc(Cl)nc1)\C(=C\[N+](=O)[O-])\NC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C11H15ClN4O2/c1-3-15(11(13-2)8-16(17)18)7-9-4-5-10(12)14-6-9/h4-6,8,13H,3,7H2,1-2H3/b11-8+
Formula Weight 270.0884
Chemical Name or Material (E)-Nitenpyram
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.