missing translation for 'onlineSavingsMsg'
Learn More

Nitinol foil, 0.05mm (0.002in) thick, superelastic, flat annealed, pickled surface

Product Code. 15493985
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15493985 25 x 25 mm Each
15403995 50 x 50 mm Each
15413995 100 x 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15493985 Supplier Thermo Scientific Alfa Aesar Supplier No. 045492.FF
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nitinol foil is used in cell-phone technology as a retractable antenna, or microphone boom, due to its highly flexible and mechanical memory nature. It is also used in some novelty products, such as self-bending spoons which can be used by amateur and stage magicians to demonstrate psychic powers or as a practical joke, as the spoon will bend itself when used to stir tea, coffee, or any other warm liquid. It is highly biocompatible and has properties suitable for use in orthopaedic implants. Due to Nitinol's unique properties it has seen a large demand for use in less invasive medical devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitinol foil
Name Note 0.05mm (0.002 in.) thick, superelastic, flat annealed, pickled surface
Quantity 25 x 25 mm
Odor Odorless
Weight ≈0.25g/25x25mm
MDL Number MFCD02091734
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.