Learn More
Nitinol foil, 0.127mm (0.005in) thick, superelastic, flat annealed, pickled surface
Beskrivelse
Innovations in the manufacturing and processing of Nitinol have led to a great expansion of Nitinol medical devices in a variety of therapeutic areas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tekniske data
Tekniske data
| Chemical Name or Material | Nitinol foil |
| Name Note | 0.127mm (0.005 in.) thick, superelastic, flat annealed, pickled surface |
| Quantity | 25 x 25 mm |
| Odor | Odorless |
| Weight | ≈0.51g/25x25mm |
| MDL Number | MFCD02091734 |
| Solubility Information | Insoluble in water. |
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.