missing translation for 'onlineSavingsMsg'
Learn More

Nitinol foil, 0.25mm (0.01in) thick, superelastic, flat annealed, pickled surface

Product Code. 11470940
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
25mm
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470940 50 x 50 mm Each
11460940 25 x 25 mm 25mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470940 Supplier Thermo Scientific Alfa Aesar Supplier No. 044953.FI
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Nitinol foil is used in photo chemically-etched parts, formed sheet shapes, EDM or laser cut shapes, cell-phone technology as a retractable antenna, or microphone boom.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitinol foil
Name Note 0.25mm (0.01 in.) thick, superelastic, flat annealed, pickled surface
Quantity 50 x 50 mm
Odor Odorless
Weight ≈1.02g/25x25mm
MDL Number MFCD02091734
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.