missing translation for 'onlineSavingsMsg'
Learn More

Nitinol tension coil spring, wire diameter 0.75mm (0.030in), mean coil diameter 5.55mm (0.22in), Alloy M

Product Code. 15484005
Change view
Click to view available options
Quantity:
100 Pc.
25 Pc.
5 Pc.
Unit Size:
Pack of 100
Pack of 25
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15484005 5 Pc. Pack of 5
15494005 25 Pc. Pack of 25
15404015 100 Pc. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15484005 Supplier Thermo Scientific Alfa Aesar Supplier No. 045655.KJ
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 7440-02-0

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitinol tension coil spring
Color Gray
Name Note Wire diameter 0.75mm, mean coil diameter 5.55mm
CAS 7440-02-0, 7440-32-6
Quantity 5 Pc.
Odor Odorless
Form Solid
MDL Number MFCD02091734
Concentration or Composition (by Analyte or Components) Nickel: 55.75%; Titanium: 44.25%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.