missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Nitrate Cuvette Tests

Product Code. p-8008130
Click to view available options
Range:
5 to 35mg/L NO3-N
Unit Size:
Pack of 25
This item is not returnable. View return policy

Product Code. 10012461

Brand: Hach Lange™ LCK340

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pre-dosed reagents for maximum safety. Easy to use and photometric evaluation, packaged in a fully equipped box. Officially approved for consent limits. Environmentally friendly due to less chemicals.

  • For use with DR3900, DR6000, DR1900, DR2800, DR3800, DR5000 instruments
  • ISO 7890-1-2-1986, DIN 38405 D9-2 certifications
  • Shelf life of 23 months from production date
  • Storage conditions of 15 - 25 °C
  • Measuring ranges of 0.23 - 13.5 mg/L NO3-N and 1.0 - 60.0 mg/L NO3
  • Precise and reliable measurement values
  • Convenient and error-free dosing of the reagents

Specifications

No. of Tests 25
Range 5 to 35mg/L NO3-N
Storage Requirements 15°C to 25°C
Test Method 2.6-Dimethylphenol
Type Nitrate Cuvette Test
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Substances and mixtures corrosive to metals Category 1
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P390-Absorb spillage to prevent material damage.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.