missing translation for 'onlineSavingsMsg'
Learn More

Nitrendipine, MedChemExpress

Product Code. 16478296
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
1g
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478296 100 mg 100mg
16474357 50 mg 50mg
16451416 200 mg 200mg
16442169 1 g 1g
16402136 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16478296 Supplier Medchem Express Supplier No. HYB0424
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nitrendipine (BAY-E-5009), an analogue of Nifedipine (HY-B0284), is a dihydropyridine calcium channel blocker with vasodilator action. Nitrendipine has antihypertensive effect.

Nitrendipine (BAY-E-5009), an analogue of Nifedipine (HY-B0284), is a dihydropyridine calcium channel blocker with vasodilator action. Nitrendipine has antihypertensive effect.

Specifications

Chemical Name or Material Nitrendipine
CAS 39562-70-4
Color Yellowish Green
Purity Grade Notes Research
Molecular Formula C18H20N2O6
Quantity 100 mg
Synonym BAY-E-5009
Solubility Information DMSO : 50 mg/mL (138.75 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=C(C)NC(C)=C(C(OC)=O)C1C2=CC=CC([N+]([O-])=O)=C2)OCC
Molecular Weight (g/mol) 360.36
Formula Weight 360.36
Percent Purity 99.25%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.