missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 2% (v/v) in Denatured Ethanol, Ricca

Product Code. 31940721
Change view
Click to view available options
Quantity:
4 L
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31940721 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 31940721 Supplier Ricca Chemical Company Supplier No. R53141004A
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
  • Nitric Acid, 2 v/v /den Alc 4 L Poly natural

Specifications

CAS 64-17-5, 67-56-1, 7697-37-2, 7732-18-5
Packaging Natural Poly Container
Quantity 4 L
Grade ACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.