missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid 70% d=1.42, for Electronic Use (MOS), Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Unit Size:
2.5 L
Specifications
Specifications
| pH | 1 |
| Color | Yellow |
| CAS | 7697-37-2 |
| Melting Point | -42°C |
| Boiling Point | 122°C |
| Packaging | HDPE plastic bottle |
| Quantity | 2.5 L |
| Molecular Formula | HNO3 |
| Physical Form | Liquid |
| MDL Number | MFCD00011349 |
| Show More |
Säkerhet och hantering
Produktbeteckning
- Nitric acid SG 1.42 (70%)
- Danger
- Oxidising liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- EUH071-Corrosive to the respiratory tract.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Nitric acid
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering