missing translation for 'onlineSavingsMsg'
Learn More

Nitric acid, Puriss. p.a., ACS Reagent, Reag. ISO, ≥69%, Honeywell Fluka

Product Code. 15603090 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
Unit Size:
2.5 L
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15603090 2.5 L Glass Bottle 2.5 L
15613090 4 x 2.5 L Glass Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15603090 Supplier Honeywell-Fluka Supplier No. 307022.5L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Puriss. p.a., ACS Reagent, Reag. ISO,≥69%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Boiling Point 122°C
CAS Min % ≥50.0000%, ≥30.0000%
CAS Max % <70.0000%, <50.0000%
Density 1.413 g/mL at 20 °C(lit.)
Packaging Glass Bottle
Quantity 2.5 L
Linear Formula HNO3
UN Number UN2031
Formula Weight 63.01
Percent Purity >69%
Grade ACS Reagent/Reagent ISO
Chemical Name or Material Nitric acid, Puriss. p.a., ACS Reagent, Reag. ISO, ≥69%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nitric acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: nitric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.