missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, Dilute, EP Grade, Reagecon™
Shop All Reagecon Products

Click to view available options
Quantity:
100 mL
Packaging:
Tamper evident
Unit Size:
100mL
Description
Nitric Acid is a corrosive mineral acid that supplies a nitro group during nitration and is also commonly used as an oxidizing agent. This solution is prepared by adding 20g nitric acid (63–70% HNO3) to 100mL water to produce a dilute solution with approximately 125g/L of HNO3.
- Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.1.1) guidelines
- Provided with a certificate of analysis and a label that shows the lot and expiration date
- Ready to use product eliminates the time and labor of preparing precise reagents and volumetric solutions
Specifications
Specifications
| Color | Colorless |
| CAS | 7697-37-2, 7732-18-5 |
| Concentration | 125g/L HNO3 |
| Packaging | Tamper evident |
| Certificate of Analysis | Yes |
| Solvent or Matrix | Water |
| Quantity | 100 mL |
| Molecular Formula | HNO3 |
| Physical Form | Liquid |
| MDL Number | MFCD00011349 |
| Show More |
Safety and Handling
Product Identifier
- NITRIC ACID, DILUTE
- Danger
- Oxidising liquid Category 2
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H272-May intensify fire; oxidiser.
- H314-Causes severe skin burns and eye damage.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : nitric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction