missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, ULTREX II Ultrapure Reagent, J.T. Baker™

Product Code. 15598374 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2 L
Packaging:
Narrow Mouth Liquid Bottle
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15598374 2 L Narrow Mouth Liquid Bottle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15598374 Supplier Avantor J.T.Baker Supplier No. 690102
  • ONLINE EXCLUSIVE PRICE
    2412.23 €
    2175.31 € / Pack of 1
    Save 236.92 €
    9% Off
 Restricted Item
Estimated Shipment: 03-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Specifications

CAS 7697-37-2
Packaging Narrow Mouth Liquid Bottle
Specific Gravity 1L = 1.41kg
Quantity 2 L
Molecular Formula HNO3
MDL Number MFCD00011349
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES O[N+]([O-])=O
IUPAC Name nitric acid
Molecular Weight (g/mol) 63.01
PubChem CID 944
ChEBI CHEBI:48107
Formula Weight 63.01
Grade ULTREX™ II Ultrapure Reagent
Chemical Name or Material Nitric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.