Learn More
Abcam Nitric Oxide Synthase Activity Assay Kit (Fluorometric)
Description
Measure NOS activity in Cell/Tissue Extracts with Nitric Oxide Synthase Activity Assay Kit AB211084. Sensitivity = 0.75 μU. Cited in 4 publications. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Quantity | 100 Tests |
| Regulatory Status | RUO |