missing translation for 'onlineSavingsMsg'
Learn More

Meditrade™ Nitril™ 3000 X-Long, Non-Sterile, Nitrile Examination Gloves

Product Code. 16650336
Change view
Click to view available options
Size:
Medium
X-Large
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16650336 X-Large Pack of 100
16640336 Medium Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 16650336 Supplier Meditrade™ Supplier No. 1282XL
  • ONLINE EXCLUSIVE PRICE
    1.90 €
    1.78 € / Pack of 100
    Save 0.12 €
    6% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Nitril™ 3000 extra long disposable nitrile examination gloves provide excellent protection for environments with a high risk of contamination. Ideal for use in clinical, laboratory, and manufacturing environments.

  • AQL 1,5
  • PPE Category III
  • Class I Medical
  • Extra long length
  • Ambidextrous
  • Non-sterile
  • Powder free and latex free
  • Textured
  • Beaded cuff

Specifications

Certifications/Compliance AQL 1,5, PPE Cat III, Class I Medical, Food compliant, EN 455-1/-2/-3/-4, EN 374-1/-2/-4/-5, EN 16623-1, EN 420, EN ISO 13495:2016, 93/42/EEC
Color Blue
Disposable Single-use
Material Nitrile
Size X-Large
Model 1282XL
Length (Metric) 29 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.