missing translation for 'onlineSavingsMsg'
Learn More

Nitril™ 3000 X-Long, Non-Sterile, Nitrile Examination Gloves

Product Code. p-7087591
Click to view available options
Size:
Medium
X-Large
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 16640336

Brand: Roesner Mautby Meditrade 1282M

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Nitril™ 3000 extra long disposable nitrile examination gloves provide excellent protection for environments with a high risk of contamination. Ideal for use in clinical, laboratory, and manufacturing environments.

  • AQL 1,5
  • PPE Category III
  • Class I Medical
  • Extra long length
  • Ambidextrous
  • Non-sterile
  • Powder free and latex free
  • Textured
  • Beaded cuff

Specifications

Certifications/Compliance AQL 1,5, PPE Cat III, Class I Medical, Food compliant, EN 455-1/-2/-3/-4, EN 374-1/-2/-4/-5, EN 16623-1, EN 420, EN ISO 13495:2016, 93/42/EEC
Color Blue
Disposable Single-use
Material Nitrile
Size Medium
Model 1282M
Length (Metric) 29 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.