missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ Nitritex™ Omega Pro-tech™ Blue Disposable Powder-free Laboratory Gloves

Product Code. 12930845
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Pack of 100
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 12930845

Brand: Nitritex™ OTEBL2105XL

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Blue disposable powder free gloves resist cuts and snags and are available in a variety of sizes.

  • Blue
  • Beaded cuff
  • Non-sterile
  • Nitrile contains no proteins
  • Various sizes
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 2859
Classification Category 3 - Complex Design Personal Protective Equipment
Color Blue
Disposable Single-use
Latex Free Yes
Material Nitrile
Product Type Nitrile Examination Glove
Size X-Large
Glove Cuff Style Beaded/Rolled
Inspected for Medical Use Yes
Length (Metric) 24 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.