Learn More
Chemsplash Jet Spray 88 Coverall Type 3B/4B 5B/6B with Feet

Description
Chemsplash Jet Spray 88 Coverall with Feet
- 88GSM Heavyweight Microporous Non Woven Fabric
- Ultrasonically Tape Welded Seams
- Self Adhesive Chin Strap for Optimum Protection
- Elasticated Hood, Half Waist, Cuffs and Ankles
- Convenient Thumb Loops at Wrist
- Single-Way Zip with Bi-Folding Self Adhesive Flap
- Silicone and Latex Free
- Non Linting Fabric
- Anti-Static
- PFAS Free
Description: Chemsplash Jet Spray 88 Coverall with Feet Type 3B/4B 5B/6B is highly resistant against chemical permeation, offering high levels of protection against even the most corrosive chemicals. Made from a thick 88gsm heavyweight polypropylene/polyethylene multilayer laminated material, it is also tested to provide protection against biological hazards to EN14126, offering protection against viruses, bacteria, and bloodborne pathogens.
Specifications
Specifications
| Product Type | Cleanroom Coveralls |
| Size | X-Large |
| Gender | Unisex |
| Material | Non-woven Fabric |
| Color | Yellow |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Attached Boots |
| Cuff Style | Elastic, Thumb Loop |
| Neck Style | Hooded |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.