missing translation for 'onlineSavingsMsg'
Learn More

Nitrilotriacetic Acid (Certified ACS), Fisher BioReagents

Product Code. 10553145 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10553145 1 kg Poly Bottle 1kg
10316843 100 g Poly Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10553145
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Nitrilotriacetic Acid is a chelating and sequestering agent. A builder in synthetic detergents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-13-9
Molecular Formula C6H9NO6
Molecular Weight (g/mol) 191.14
MDL Number MFCD00004287
InChI Key MGFYIUFZLHCRTH-UHFFFAOYSA-N
Synonym nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i
PubChem CID 8758
ChEBI CHEBI:44557
IUPAC Name 2-[bis(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CC(O)=O)CC(O)=O

Specifications

Melting Point 245°C
Color White
pH 2.3
Assay Percent Range ≥98.0 %
Packaging Poly Bottle
Linear Formula N(CH2COOH)3
Quantity 1 kg
Merck Index 15, 6665
Formula Weight 191.14
Percent Purity ≥98.0%
Grade Certified ACS
Physical Form Powder or Solid
Chemical Name or Material Nitrilotriacetic Acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nitrilotriacetic acid
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.