missing translation for 'onlineSavingsMsg'
Learn More

Nitrilotriacetic acid, Solstice

Product Code. 15684240 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 g
6 x 50 g
25 kg
Unit Size:
25kg
50g
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15684240 25 kg 25kg
15694240 50 g 50g
15624250 6 x 50 g Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15684240 Supplier Honeywell-Fluka Supplier No. 3453925KG
  • ONLINE EXCLUSIVE PRICE
    3885.00 €
    3652.93 € / 25kg
    Save 232.07 €
    5% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for complexometry,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-13-9
Molecular Formula C6H9NO6
Molecular Weight (g/mol) 191.14
MDL Number MFCD00004287
InChI Key MGFYIUFZLHCRTH-UHFFFAOYSA-N
Synonym nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i
PubChem CID 8758
ChEBI CHEBI:44557
IUPAC Name 2-[bis(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CC(O)=O)CC(O)=O

Specifications

Flash Point 100°C (212°F)
Linear Formula N(CH2COOH)3
Quantity 25 kg
UN Number NONH for all modes of transport
Beilstein 1710776
Formula Weight 191.14g/mol
Percent Purity ≥99%
Chemical Name or Material Nitrilotriacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.