missing translation for 'onlineSavingsMsg'
Learn More

Nitrilotriacetic Acid, TRC

Product Code. 17971915
Change view
Click to view available options
Quantity:
1 g
10 g
250 g
Unit Size:
10g
1g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17971915 250 g 250g
17934245 1 g 1g
17902935 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17971915 Supplier Toronto Research Chemicals Supplier No. TRCN490175250G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-13-9
Molecular Formula C6 H9 N O6
Molecular Weight (g/mol) 191.14
Synonym Nitrilotriacetic acid, 2-[Bis(carboxymethyl)amino]acetic acid, NTA, Glycine, N,N-bis(carboxymethyl)-, Acetic acid, nitrilotri- (8CI), N,N-Bis(carboxymethyl)glycine, Chel 300, Chelest NT, Complexon I, Dissolvine A, Dissolvine AZ, Hampshire NTA acid, N 0098, NSC 2121, NTA, Nitrilo-2,2',2' '-triacetic acid, Nitrilo-N,N,N-triacetic acid, Nitrilotris(methylenecarboxylic acid), Titriplex I, Triglycine, Triglycollamic acid, Trilon AS, Tris(carboxymethyl)amine, Tris(hydroxycarbonylmethyl)amine, Versene NTA acid, α,α',α' '-Trimethylaminetricarboxylic acid
IUPAC Name 2-[bis(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CC(=O)O)CC(=O)O

Specifications

InChI Formula InChI=1S/C6H9NO6/c8-4(9)1-7(2-5(10)11)3-6(12)13/h1-3H2,(H,8,9)(H,10,11)(H,12,13)
Quantity 250 g
Formula Weight 191.043
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Nitrilotriacetic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.