missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Luminescence Enhancer For Membrane Blotting Assays, Nitro-Block-II™ Enhancer

Product Code. 12604335 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
20 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12604335 20 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12604335 Supplier Invitrogen™ Supplier No. T2184
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Use with CSPD™ or CDP-Star™ chemiluminescent substrates to increase the alkaline phosphatase signal for protein or nucleic acid blots. The Nitro-Block-II membrane enhancer increases chemiluminescent signal intensity on nitrocellulose and PVDF

This concentrated Nitro-Block-II™ Enhancer can be used with CSPD™ or CDP-Star™ chemiluminescent substrates to increase the alkaline phosphatase signal for protein or nucleic acid blots.

The Nitro-Block-II™ membrane enhancer increases chemiluminescent signal intensity on nitrocellulose and PVDF membranes.

  • Chemically optimized formulations provide enhanced signal intensity in blotting applications with dioxetane-based substrate solutions (CSPD™ or CDP-Star™ substrates) on nitrocellulose or PVDF membranes.
  • Enhancer increases signal intensity, allowing decreased signal acquisition time.

Increased Signal, Shorter Exposures

Without enhancer, the chemiluminescent signal on nitrocellulose membranes is very weak, making detection either impossible or very time consuming. With enhancer, short exposures (10 to 45 minutes) can be performed on the same material. Although PVDF membranes do not require the use of Nitro-Block-II™ enhancer, exposure times are ten-fold faster with enhancer. This permits very short exposure times on PVDF, ranging from 15 seconds to 15 minutes for western blots. Nitro-Block-II™ enhancer may be used with CSPD™ substrate on nitrocellulose or PVDF membranes. Nitro-Block-II™ enhancer should only be used with CDP-Star™ substrate on nitrocellulose membranes.

For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Luminescence Enhancer
Content And Storage Store in refrigerator (2°C to 8°C).
Format Bottle(s)
Form Liquid
Shipping Condition Room Temperature
Substrate Type AP (Alkaline Phosphatase) Substrate
Substrate Alkaline Phosphatase
Product Line Nitro-Block-ll, NovaBright
Quantity 20 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.