missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Nitro blue tetrazolium chloride

Product Code. 16461796
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461796 500 mg 500mg
16439865 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16461796 Supplier MedChemExpress Supplier No. HY15925
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nitro blue tetrazolium chloride (NBT) is a substrate for dehydrogenases; is used with the alkaline phosphatase substrate 5-Bromo-4-Chloro-3-Indolyl Phosphate (BCIP) in western blotting and immunohistological staining procedures.

Specifications

Quantity 500 mg
Storage Requirements 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Solubility 70%DMF : 100 mg/mL (122.30 mM; Need ultrasonic) ∣ DMSO : 33.33 mg/mL (40.76 mM; Need ultrasonic) ∣ H2O : 8 mg/mL (9.78 mM; Need ultrasonic)
Shipping Condition Room temperature
CAS 298-83-9
Molecular Weight (g/mol) 817.64 g/mol
Synonym NBT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.