missing translation for 'onlineSavingsMsg'
Learn More

Nitro Blue Tetrazolium, Fisher BioReagents

Product Code. 10679933
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10679933 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10679933 Supplier Fisher BioReagents Supplier No. BP1081

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used in conjunction with BCIP as a substrate for acid and alkaline phosphatases

Nitro BT is used in conjunction with BCIP as a substrate for acid and alkaline phosphatases.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 189°C
CAS 298-83-9
Extinction Coefficient 650g(-1) cm(-1) min.
Molecular Formula C40H30Cl2N10O6
Synonym NBT, Nitro BT, p-Nitro Blue Tetrazolium
Identification Pass Test
InChI Key FSVCQIDHPKZJSO-UHFFFAOYSA-L
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]
IUPAC Name 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride
Molecular Weight (g/mol) 817.644
PubChem CID 9281
ChEBI CHEBI:9505
Lambda Max 258 to 262nm
Formula Weight 817.65
Packaging Amber Glass
Color Yellow
Quantity 1 g
Physical Form Powder
Chemical Name or Material Nitro Blue Tetrazolium
Show More Show Less
compliance-icons
Product Identifier
  • Nitro blue tetrazolium
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.