missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nitrocellulose Membranes, 0.2 μm

Product Code. 10096363 Shop All Thermo Scientific Products
Change view
Click to view available options
Dimensions (LxW):
10.5 x 7.9 cm
12 x 8 cm
8 x 8 cm
Sufficient For:
15 Mini-gel Blots
25 Mini-gel Blots
Unit Size:
0.25kg
Pack of 15
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (LxW) Sufficient For Unit Size
10096363 8 x 8 cm 15 Mini-gel Blots Pack of 15
10148113 10.5 x 7.9 cm 15 Mini-gel Blots Pack of 15
10561674 12 x 8 cm 25 Mini-gel Blots 0.25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10096363 Supplier Thermo Scientific™ Supplier No. 88024
Protein Biology Essentials
Use promo code 27946
View all promotions
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Nitrocellulose is a popular binding matrix for western blotting because of its high affinity for proteins and compatibility with a variety of detection methods (western blotting, dot-blot assays, and other protein or nucleic acid methods).

Nitrocellulose is a popular binding matrix for western blotting because of its high affinity for proteins and compatibility with a variety of detection methods (western blotting, dot-blot assays, and other protein or nucleic acid methods). The 0.2-μm pore size is well-suited for western blotting, and this smaller pore size is ideal for the transfer of low molecular weight proteins or peptides and nucleic acids (<300 bp).

Features include:
100% pure nitrocellulose membrane
• Convenient—save time with pre-cut sheets
• Compatible with commonly used transfer conditions and detection methods such as staining, immunodetection, chemiluminescence, fluorescence, and radiolabeling
• High-binding affinity—provides excellent protein binding, blocks easily, and does not require pre-wetting with alcohol
• Low background—produce high-quality data with low background signal in both chemiluminescent and fluorescent western blotting

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 8 cm
Width (Metric) 8 cm
Description Nitrocellulose Membranes, 0.2 μm, 8 x 8 cm
Binding Capacity ∼209 μg/cm2 of protein
Quantity 15 Pre-cut Membranes
Format Sheet
Material Nitrocellulose
Pore Size 0.2 μm
Content And Storage Store membranes flat at ambient temperature, away from chemical vapors. Some solvent vapors may partially dissolve the membranes, which will disrupt the pore structure. Keep membranes out of direct sunlight. Sunlight may cause the nitrocellulose to dry and become brittle.
Note: Nitrocellulose membrane filters are highly flammable. Keep away from heat and open flame. Flash point is approximately 2°C. Membranes can be sterilized by steam-autoclaving at 121°C.
Shipping Condition Room Temperature
Dimensions (LxW) 8 x 8 cm
Sufficient For 15 Mini-gel Blots
Show More Show Less
compliance-icons
Product Identifier
  • Nitrocellulose Membranes, 0.2 µm, 8 x 8 cm
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: nitrocellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.