missing translation for 'onlineSavingsMsg'
Learn More

Melford Nitrofurantoin

Product Code. 30018592
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018592 5 g 5g
30018589 1 g 1g
30018591 25 g 25g
30018590 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30018592 Supplier Melford Supplier No. N515005.0
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nitrofurantoin works by inhibiting bacterial enzymes involved in the metabolism of certain compounds, leading to the disruption of bacterial DNA, RNA, and protein synthesis.

Nitrofurantoin works by inhibiting bacterial enzymes involved in the metabolism of certain compounds, leading to the disruption of bacterial DNA, RNA, and protein synthesis.

Researchers may use nitrofurantoin as a selective agent by incorporating it into culture media to promote the growth of cells or organisms that have been genetically engineered to express resistance to this antibiotic. It may have applications in microbiology and cell culture experiments, particularly in the context of antibiotic selection.

Specifications

CAS 67-20-9
Molecular Formula C8H6N4O5
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.