missing translation for 'onlineSavingsMsg'
Learn More

Nitrofurantoin, 98%, Thermo Scientific™

45.90 € - 574.00 €

Chemical Identifiers

CAS 67-20-9
Molecular Formula C8H6N4O5
Molecular Weight (g/mol) 238.16
MDL Number MFCD00003224
InChI Key NXFQHRVNIOXGAQ-YCRREMRBSA-N
Synonym nitrofurantoin, macrodantin, 5-nitrofurantoin, furadantin, furadonine, nitrofurantoine, furadantine, furadantoin, furadoine, furadontin
PubChem CID 6604200
ChEBI CHEBI:71415
IUPAC Name 1-[(E)-[(5-nitrofuran-2-yl)methylidene]amino]imidazolidine-2,4-dione
SMILES [O-][N+](=O)C1=CC=C(O1)\C=N\N1CC(=O)NC1=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11421077
View Documents
Thermo Scientific Alfa Aesar
B24079.14
25g
45.90 €
25g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
11431077
View Documents
Thermo Scientific Alfa Aesar
B24079.22
100g
136.00 €
100g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
11441077
View Documents
Thermo Scientific Alfa Aesar
B24079.36
500g
574.00 €
500g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Nitrofurantoin is a nitrofuran antibiotic that is used as a substrate of bacterial nitrofuran reductase to study the interactions of active metabolites with DNA, ribosomal proteins and metabolic and pro-oxidant processes. It is an antibactericidal compound that has been historically prepared by the reaction of 1-aminohydantoin sulfate and 5-nitro-2-furaldehyde diacetate. It shows activity against many Gram-positive and Gram-negative bacteria. Nitrofurantoin is effective against enterococci, staphylococci, streptococci, corneybacteria, many strains of Escherichia coli.

Solubility
Soluble in water (<0.01g/100mL at 19°C), dimethyl sulfoxide and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature. Incompatible with oxidizing agents. It is sensitive to light.
Specifications

Chemical Identifiers

67-20-9
238.16
NXFQHRVNIOXGAQ-YCRREMRBSA-N
6604200
1-[(E)-[(5-nitrofuran-2-yl)methylidene]amino]imidazolidine-2,4-dione
C8H6N4O5
MFCD00003224
nitrofurantoin, macrodantin, 5-nitrofurantoin, furadantin, furadonine, nitrofurantoine, furadantine, furadantoin, furadoine, furadontin
CHEBI:71415
[O-][N+](=O)C1=CC=C(O1)\C=N\N1CC(=O)NC1=O

Specifications

67-20-9
MFCD00003224
nitrofurantoin, macrodantin, 5-nitrofurantoin, furadantin, furadonine, nitrofurantoine, furadantine, furadantoin, furadoine, furadontin
NXFQHRVNIOXGAQ-YCRREMRBSA-N
1-[(E)-[(5-nitrofuran-2-yl)methylidene]amino]imidazolidine-2,4-dione
6604200
238.16
Nitrofurantoin
C8H6N4O5
Hygroscopic; Light sensitive
Soluble in water (<0.01g/100 ml at 19°C),dimethyl sulfoxide and methanol.
[O-][N+](=O)C1=CC=C(O1)\C=N\N1CC(=O)NC1=O
238.16
CHEBI:71415
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H334-H341-H351-H361-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
May cause an allergic skin reaction.

P260-P264b-P270-P272-P280g-P285-P301+P312-P302+P352-P304+P341-P314-P330-P333+P313-P342+P311-P363-P501c

H302-H317-H334-H373

missing translation for 'einecsNumber' : 200-646-5

missing translation for 'rtecsNumber' : MU2800000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Product Identifier
  • Nitrofurantoin

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contain: Nitrofurantoin

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.