missing translation for 'onlineSavingsMsg'
Learn More

Nitrofurantoin, 98%

Product Code. 11926821 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11926821 10 g 10g
11946821 25 g 25g
11936821 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11926821 Supplier Thermo Scientific Acros Supplier No. 229380100
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 67-20-9 | C8H6N4O5 | 238.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-20-9
Molecular Formula C8H6N4O5
Molecular Weight (g/mol) 238.16
MDL Number MFCD00003224
InChI Key NXFQHRVNIOXGAQ-YCRREMRBSA-N
Synonym nitrofurantoin, macrodantin, 5-nitrofurantoin, furadantin, furadonine, nitrofurantoine, furadantine, furadantoin, furadoine, furadontin
PubChem CID 6604200
ChEBI CHEBI:71415
SMILES [O-][N+](=O)C1=CC=C(O1)\C=N\N1CC(=O)NC1=O

Specifications

Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 10 g
IUPAC Name 1-[(E)-(5-nitrofuran-2-yl)methylideneamino]imidazolidine-2,4-dione
Formula Weight 238.16
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material Nitrofurantoin
compliance-icons compliance-icons
Product Identifier
  • Nitrofurantoin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.